o-Carborane ≥95%

Supplier: STREM CHEMICALS INC MS

05-0300-5G 05-0300-1G
100197-292EA 680.8 USD
100197-292 100197-290
o-Carborane ≥95%
o-Carborane
Formula: C₂H₁₂B₁₀
MW: 144.23 g/mol
Melting Pt: 285…287 °C
MDL Number: MFCD00012169
CAS Number: 16872-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR