Borane-tert-butylamine complex ≥97%, Powder

Supplier: STREM CHEMICALS INC MS
Danger

05-0101-25G 05-0101-100G
100204-770EA 108.98 USD
100204-770 100209-494
Borane-tert-butylamine complex ≥97%, Powder
Borane tert-butylamine complex
Formula: C₄H₁₄BN
Melting Pt: 98 °C (dec.)
MDL Number: MFCD00075635
CAS Number: 7337-45-3
UN: 2926
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR