Bis(neopentyl glycolato)diboron ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane

05-0025-1G 05-0025-5G
100204-466EA 53 USD
100204-466 100204-468
Bis(neopentyl glycolato)diboron ≥97%
Bis(neopentyl glycolato)diboron

Boronic Acid and Ester

Formula: C₁₀H₂₀B₂O₄
Melting Pt: 182…185 °C
MDL Number: MFCD02093062
CAS Number: 201733-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR