Bis(hexylene glycolato)diboron ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)

05-0020-1G 05-0020-5G
100204-462EA 118.89 USD
100204-462 100204-464
Bis(hexylene glycolato)diboron ≥99%
Bis(hexylene glycolato)diboron

Boronic Acid and Ester

Formula: C₁₂H₂₄B₂O₄
Melting Pt: 98…101 °C
MDL Number: MFCD06246008
CAS Number: 230299-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR