(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium
03-5001-1G
03-5001-5G
03-5001-25G
100207-512EA
66.52
USD
100207-512
100207-514
100206-184
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium
Metal TMHD, Volatile Precursors for CVD
Formula:
C₁₁H₁₉LiO₂ MW: 190.24 g/mol Boiling Pt: 295 °C (dec.) Melting Pt: 265…268 °C |
MDL Number:
MFCD00142754 CAS Number: 22441-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...