Lithium trifluoroacetate monohydrate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
03-5000-25G
03-5000-100G
100208-744EA
100.49
USD
100208-744
100208-742
Lithium trifluoroacetate monohydrate ≥97%
Lithium trifluoroacetate monohydrate
Formula:
C₂F₃LiO₂·H₂O MW: 137.97 g/mol (119.96 anhy) |
MDL Number:
MFCD00149663 CAS Number: 2923-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...