Lithium tri-tert-butoxyaluminum hydride ~1.0M in THF

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lithium tri-tert-butoxyaluminohydride

03-4850-500MMOL 03-4850-100MMOL
100203-890EA 1220.06 USD
100203-890 100203-888
Lithium tri-tert-butoxyaluminum hydride ~1.0M in THF
Lithium tri-tert-butoxyaluminum hydride
Formula: C₁₂H₂₈AlLiO₃
MW: 254.27 g/mol
Boiling Pt: 66 °C (THF)
Melting Pt: –108° (THF)
MDL Number: MFCD00011532
CAS Number: 17476-04-9
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR