Lithium tris(S-(-)-1,1'-binaphthyl-2,2'-diolato)yttrate(III) tetrahydrofuran adduct ≥97%

Supplier: STREM CHEMICALS INC MS

03-2010-250MG 03-2010-1G
101324-490EA 444.43 USD
101324-490 101324-492
Lithium tris(S-(-)-1,1'-binaphthyl-2,2'-diolato)yttrate(III) tetrahydrofuran adduct ≥97%
Lithium tris(S-(-)-1,1'-binaphthyl-2,2'-diolato)yttrate(III) tetrahydrofuran adduct
MDL Number: MFCD11112164
CAS Number: 500995-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR