Lithium trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

03-2000-10G 03-2000-50G
100197-286EA 69.35 USD
100197-286 100205-812
Lithium trifluoromethanesulfonate ≥99%
Lithium trifluoromethanesulphonate

Metal Triflate

Formula: CF₃LiO₃S
MW: 156.01 g/mol
MDL Number: MFCD00013227
CAS Number: 33454-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR