Lithium pentamethylcyclopentadienide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

03-1150-25G 03-1150-5G 03-1150-1G
100199-690EA 1142.22 USD
100199-690 100199-692 100199-688
Lithium pentamethylcyclopentadienide ≥98%
Lithium pentamethylcyclopentadienide

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00269823
CAS Number: 51905-34-1
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR