Tetrabutylammonium fluoride hydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

02-5015-25G 02-5015-5G
100210-718EA 389.23 USD
100210-718 100203-882
Tetrabutylammonium fluoride hydrate ≥98%
Tetrabutylammonium fluoride hydrate
Formula: C₁₆H₃₆FN·H₂O
MDL Number: MFCD00149980
CAS Number: 22206-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR