Ammonium tetrachloroaurate hydrate ≥99.9985% (Au basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ammonium tetrachloroaurate(III) hydrate

02-1000-1G 02-1000-5G
100202-832EA 284.49 USD
100202-832 100199-216
Ammonium tetrachloroaurate hydrate ≥99.9985% (Au basis), PURATREM
Ammonium tetrachloroaurate hydrate
Formula: H₄AuCl₄N·H₂O
MW: 356.82 g/mol (anhy)
MDL Number: MFCD00011503
CAS Number: 13874-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR