Ammonium hexachlororhodate(III) monohydrate ≥99.995% (Rh basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
02-0590-5G
02-0590-1G
100206-070EA
3020.43
USD
100206-070
100207-506
Ammonium hexachlororhodate(III) monohydrate ≥99.995% (Rh basis), PURATREM
Ammonium hexachlororhodate(III) monohydrate
Formula:
H₁₂Cl₆N₃Rh·H₂O MW: 369.74 g/mol (anhy) |
MDL Number:
MFCD00003394 CAS Number: 15336-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...