Benzyl (3S)-3-hydroxypiperidine-1-carboxylate ≥99.9%

Synonyms: (S)-1-Cbz-3-hydroxypiperidine

00792-1 00792-2 00792-3
103534-450EA 30.77 USD
103534-450 103534-452 103534-454
Benzyl (3S)-3-hydroxypiperidine-1-carboxylate ≥99.9%
Benzyl (3S)-3-hydroxypiperidine-1-carboxylate
Formula: C₁₃H₁₇NO₃
MW: 235.279 g/mol
MDL Number: MFCD11112281
CAS Number: 94944-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR