Benzyl-3-oxopiperidine-1-carboxylate

Synonyms: 1-Benzyloxycarbonyl-3-piperidone, Benzyl 3-oxopiperidine-1-carboxylate

00786-1 00786-2 00786-3
103534-444EA 30.77 USD
103534-444 103534-446 103534-448
Benzyl-3-oxopiperidine-1-carboxylate
Benzyl-3-oxopiperidine-1-carboxylate
Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
MDL Number: MFCD03001723
CAS Number: 61995-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR