Benzyl-4-oxo-1-piperidinecarboxylate ≥99.4%

Synonyms: 1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine

00785-1 00785-2 00785-3
103534-438EA 30.77 USD
103534-438 103534-440 103534-442
Benzyl-4-oxo-1-piperidinecarboxylate ≥99.4%
Benzyl-4-oxo-1-piperidinecarboxylate
Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
MDL Number: MFCD00673144
CAS Number: 19099-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR