4-(Boc-amino)piperidine ≥97%

Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

00010-1 00010-2 00010-3
103534-420EA 12.31 USD
103534-420 103534-422 103534-424
4-(Boc-amino)piperidine ≥97%
4-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD00798171
CAS Number: 73874-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR