Sodium bromide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals

02119-2.5KG 02119-500G 02119-1KG
BJ02119-2.5KGEA 229.76 USD
BJ02119-2.5KG BJ02119-500G BJ-02119-1KG
Sodium bromide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396
Melting Pt: 753
Density: 3.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Assay (calculated to the dried substance) 98.5 - 100.5 %
Loss on drying (105°C, 3h) Max. 1.0 %
Acidity or alkalinity complying
Arsenic (As) Max. 0.0002 %
Barium (Ba) Max. 0.002 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 10 ppm
Magnesium (Mg) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Bromate (BrO₃) complying
Chloride (Cl) Max. 0.2 %
Iodide (I) complying
Sulfate (SO₄) Max. 0.005 %
Mg, earth alkali metals (as Ca) Max. 200 ppm
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR