Human Pfn2 Cg75-1Mg, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CG75-1MG CG75-500UG CG75-50UG CG75-10UG
102499-274EA 1809.23 USD
102499-274 102499-276 102499-278 102509-534
Human Pfn2 Cg75-1Mg, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Profilin-2 is produced by our E.coli expression system and the target gene encoding Met1-Phe140 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.