Human Recombinant TIM-3
Supplier: Bon Opus Biosciences
CU05-50UG CU05-10UG
103529-264EA 254.15 USD
103529-264 103529-338 103529-396 103529-414
Human Recombinant TIM-3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human T-Cell Membrane Protein 3 is produced by our Mammalian expression system and the target gene encoding Ser22-Arg200 is expressed with a Fc & Avi tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.