Recombinant Signal Transducer CD24
Supplier: Bon Opus Biosciences
CS99-10UG CS99-50UG CS99-500UG CS99-1MG
103529-212EA 85.08 USD
103529-212 103529-254 103529-328 103529-538
Recombinant Signal Transducer CD24
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Signal Transducer CD24 is produced by our Mammalian expression system and the target gene encoding Ser27-Gly59 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.