Recombinant Myeloperoxidase
Supplier: Bon Opus Biosciences
CS87-10UG CS87-1MG CS87-50UG CS87-500UG
103529-188EA 118.46 USD
103529-188 103529-378 103529-462 103529-504
Recombinant Myeloperoxidase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Myeloperoxidase is produced by our Mammalian expression system and the target gene encoding Met16-Thr718 is expressed with a 10His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.