Recombinant Decorin
Supplier: Bon Opus Biosciences
CS86-10UG CS86-1MG CS86-50UG CS86-500UG
103529-186EA 57.08 USD
103529-186 103529-376 103529-460 103529-502
Recombinant Decorin
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Decorin is produced by our Baculovirus expression system and the target gene encoding Gly17-Lys359 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.