Mouse Recombinant Protein S100-A4
Supplier: Bon Opus Biosciences
CR30-10UG CR30-1MG CR30-50UG CR30-500UG
103529-182EA 85.08 USD
103529-182 103529-372 103529-456 103529-498
Mouse Recombinant Protein S100-A4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse S100A4 is produced by our E.coli expression system and the target gene encoding Met1-Lys101 is expressed with a 6His at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.