Mouse Recombinant Interleukin-1 Receptor-like 1
Supplier: Bon Opus Biosciences
CK89-10UG CK89-1MG CK89-50UG CK89-500UG
102970-244EA 107.69 USD
102970-244 102970-262 102970-362 102970-456
Mouse Recombinant Interleukin-1 Receptor-like 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Interleukin-1 receptor-like 1 is produced by our Mammalian expression system and the target gene encoding Ser27-Ala337 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.