Mouseb7-1/Cd80, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CK87-10UG CK87-50UG CK87-500UG CK87-1MG
102970-240EA 34.46 USD
102970-240 102970-358 102970-452 102970-546
Mouseb7-1/Cd80, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Activation B7-1 antigen is produced by our Mammalian expression system and the target gene encoding Val38-Lys245 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.