Mouse Recombinant Interferon Alpha-2
Supplier: Bon Opus Biosciences
CK83-10UG CK83-50UG CK83-500UG CK83-1MG
102970-232EA 34.46 USD
102970-232 102970-350 102970-444 102970-538
Mouse Recombinant Interferon Alpha-2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Interferon alpha-2 is produced by our E.coli expression system and the target gene encoding Cys24-Glu190 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.