Mouseb7-2/Cd86, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CK79-10UG CK79-50UG CK79-500UG CK79-1MG
102970-224EA 34.46 USD
102970-224 102970-342 102970-436 102970-530
Mouseb7-2/Cd86, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse T-lymphocyte activation antigen CD86 is produced by our Mammalian expression system and the target gene encoding Val26-Glu245 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.