Mouse Recombinant Insulin-like Growth Factor-Binding Protein 6
Supplier: Bon Opus Biosciences
CK73-10UG CK73-50UG CK73-500UG CK73-1MG
102970-208EA 118.46 USD
102970-208 102970-330 102970-424 102970-518
Mouse Recombinant Insulin-like Growth Factor-Binding Protein 6
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Insulin-Like Growth Factor-Binding Protein 6 is produced by our Mammalian expression system and the target gene encoding Ala26-Gly238 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.