Humanox40/Tnfrsf4ck67
Supplier: Bon Opus Biosciences
CK67-10UG CK67-50UG CK67-500UG CK67-1MG
102970-192EA 102.31 USD
102970-192 102970-318 102970-412 102970-506
Humanox40/Tnfrsf4ck67
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant Mouse Interleukin-2 receptor subunit alpha is produced by our Mammalian expression system and the target gene encoding Glu22-Lys236 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.