Human Recombinant Tumor necrosis factor Receptor superfamily member 4
Supplier: Bon Opus Biosciences
CK60-10UG CK60-50UG CK60-500UG CK60-1MG
102970-280EA 107.69 USD
102970-280 102970-302 102970-396 102970-490
Human Recombinant Tumor necrosis factor Receptor superfamily member 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human OX40 is produced by our Mammalian expression system and the target gene encoding Leu29-Ala216 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.