Mouse Interleukin-16/Il-16, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CK11-10UG CK11-50UG CK11-500UG CK11-1MG
102942-346EA 118.46 USD
102942-346 102942-348 102942-350 102942-352
Mouse Interleukin-16/Il-16, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Interleukin-16 is produced by our E.coli expression system and the target gene encoding Ser1205-Ser1322 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.