Mouse Recombinant Tumor necrosis Factor Ligand Superfamily Member 11
Supplier: Bon Opus Biosciences
CJ94-10UG CJ94-50UG CJ94-500UG CJ94-1MG
102944-930EA 170.15 USD
102944-930 102944-932 102944-934 102944-936
Mouse Recombinant Tumor necrosis Factor Ligand Superfamily Member 11
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse TNF-related Activation-induced Cytokine is produced by our Mammalian expression system and the target gene encoding Ala73-Asp316 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.