Mouse Recombinant Programmed Cell Death 1 Ligand 1Cd274
Supplier: Bon Opus Biosciences
CJ89-10UG CJ89-50UG CJ89-500UG CJ89-1MG
102944-890EA 57.08 USD
102944-890 102944-892 102944-894 102944-896
Mouse Recombinant Programmed Cell Death 1 Ligand 1Cd274
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Programmed Cell Death 1 Ligand 1 is produced by our Mammalian expression system and the target gene encoding Phe19-Thr238 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.