Human Recombinant Interleukin-2 Receptor Subunit alpha
Supplier: Bon Opus Biosciences
CJ78-10UG CJ78-50UG CJ78-500UG CJ78-1MG
102944-802EA 102.31 USD
102944-802 102944-804 102944-806 102944-808
Human Recombinant Interleukin-2 Receptor Subunit alpha
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human IL-2 receptor alpha is produced by our Mammalian expression system and the target gene encoding Glu22-Cys213 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.