Human Recombinant Ephrin-A5
Supplier: Bon Opus Biosciences
CJ76-10UG CJ76-50UG CJ76-500UG CJ76-1MG
102944-786EA 118.46 USD
102944-786 102944-788 102944-790 102944-792
Human Recombinant Ephrin-A5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Ephrin-A5 is produced by our Mammalian expression system and the target gene encoding Gln21-Asn203 is expressed with a Fc tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.