Mouse Recombinant Ephrin-B1
Supplier: Bon Opus Biosciences
CJ23-10UG CJ23-50UG CJ23-500UG CJ23-1MG
102944-386EA 34.46 USD
102944-386 102944-388 102944-390 102944-392
Mouse Recombinant Ephrin-B1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Ephrin-B1 is produced by our Mammalian expression system and the target gene encoding Lys30-Ser229 is expressed with a Fc, 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.