Recombinant Human Chemerin, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CJ21-10UG CJ21-50UG CJ21-500UG CJ21-1MG
102944-370EA 118.46 USD
102944-370 102944-372 102944-374 102944-376
Recombinant Human Chemerin, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Chemerin is produced by our Mammalian expression system and the target gene encoding Glu21-Ser157 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.