Human Recombinant SPINK7/ECRG-2/Esophagus cancer-related gene 2 protein/Serine protease inhibitor Kazal-type 7/ECG2
Supplier: Bon Opus Biosciences
CJ18-10UG CJ18-50UG CJ18-500UG CJ18-1MG
102944-346EA 170.15 USD
102944-346 102944-348 102944-350 102944-352
Human Recombinant SPINK7/ECRG-2/Esophagus cancer-related gene 2 protein/Serine protease inhibitor Kazal-type 7/ECG2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SPINK7 is produced by our Mammalian expression system and the target gene encoding Ser20-Cys85 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.