Mouse Recombinant CNDP2/CNDP dipeptidase 2/Cytosolic non-specific dipeptidase/Glutamate carboxypeptidase-like protein 1/Cn2
Supplier: Bon Opus Biosciences
CJ14-10UG CJ14-50UG CJ14-500UG CJ14-1MG
102944-314EA 118.46 USD
102944-314 102944-316 102944-318 102944-320
Mouse Recombinant CNDP2/CNDP dipeptidase 2/Cytosolic non-specific dipeptidase/Glutamate carboxypeptidase-like protein 1/Cn2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse CNDP dipeptidase 2 is produced by our Mammalian expression system and the target gene encoding Met1-Asn475 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.