Human Recombinant Myelin protein zero-like 1
Supplier: Bon Opus Biosciences
CI85-10UG CI85-50UG CI85-500UG CI85-1MG
102944-106EA 57.08 USD
102944-106 102944-108 102944-110 102944-112
Human Recombinant Myelin protein zero-like 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Myelin Protein P0-like 1 is produced by our Mammalian expression system and the target gene encoding Ser36-Val162 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.