Human Recombinant Pancreatic prohormone
Supplier: Bon Opus Biosciences
CI10-10UG CI10-1MG CI10-500UG CI10-50UG
102499-702EA 170.15 USD
102499-702 102499-704 102499-706 102499-708
Human Recombinant Pancreatic prohormone
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Pancreatic Polypeptide is produced by our Mammalian expression system and the target gene encoding Ala30-Arg88 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.