Recombinant Human Neuregulin-1 beta, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CH86-10UG CH86-50UG CH86-500UG CH86-1MG
102943-722EA 118.46 USD
102943-722 102943-724 102943-726 102943-728
Recombinant Human Neuregulin-1 beta, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Neuregulin-1 beta is produced by our E.coli expression system and the target gene encoding Ser177-Glu241 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.