Human Recombinant Microtubule-Associated Protein tau
Supplier: Bon Opus Biosciences
CH19-10UG CH19-1MG CH19-500UG CH19-50UG
102499-508EA 118.46 USD
102499-508 102499-510 102499-512 102499-514
Human Recombinant Microtubule-Associated Protein tau
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Microtubule-Associated Protein Tau-F is produced by our E.coli expression system and the target gene encoding Met1-Leu441 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.