Human Recombinant Fibroblast Growth Factor 19
Supplier: Bon Opus Biosciences
CG74-10UG CG74-1MG CG74-500UG CG74-50UG
102509-526EA 118.46 USD
102509-526 102509-528 102509-530 102509-532
Human Recombinant Fibroblast Growth Factor 19
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Fibroblast growth factor 19 is produced by our E.coli expression system and the target gene encoding Phe27-Lys216 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.