Human Recombinant Rheb
Supplier: Bon Opus Biosciences
CG22-10UG CG22-1MG CG22-500UG CG22-50UG
102865-238EA 170.15 USD
102865-238 102865-240 102865-242 102865-244
Human Recombinant Rheb
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human GTP-binding protein Rheb is produced by our E.coli expression system and the target gene encoding Met1-Met184 is expressed with a GST tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.