Human Recombinant Sulfotransferase 1A1
Supplier: Bon Opus Biosciences
CF97-10UG CF97-1MG CF97-500UG CF97-50UG
102865-078EA 170.15 USD
102865-078 102865-080 102865-082 102865-084
Human Recombinant Sulfotransferase 1A1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Sulfotransferase 1A1 is produced by our E.coli expression system and the target gene encoding Met1-Leu295 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.