Recombinant Human Vcp, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CF48-10UG CF48-1MG CF48-500UG CF48-50UG
102864-798EA 170.15 USD
102864-798 102864-800 102864-802 102864-804
Recombinant Human Vcp, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Valosin-Containing Protein is produced by our E.coli expression system and the target gene encoding Ala2-Asn589 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.