Recombinant Human Aldh1A1, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CF44-10UG CF44-1MG CF44-500UG CF44-50UG
102864-774EA 107.69 USD
102864-774 102864-776 102864-778 102864-780
Recombinant Human Aldh1A1, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Aldehyde Dehydrogenase 1-A1 is produced by our E.coli expression system and the target gene encoding Met1-Ser501 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.