Human Recombinant Aif1
Supplier: Bon Opus Biosciences
CF31-10UG CF31-1MG CF31-500UG CF31-50UG
102864-694EA 118.46 USD
102864-694 102864-696 102864-698 102864-700
Human Recombinant Aif1
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human allograft inflammatory factor 1 is produced by our E.coli expression system and the target gene encoding Ser2-Pro147 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.